Learn More
Genscript Corporation dNTP 10 mM each 99 HPLC
Catalog No. 5000058944
Encompass
10 mM dNTP Mix is suitable for use in PCR sequencing nick translation fill-ins cDNA synthesis TdT tailing reactions and the dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in sterilized water The 10 mM dNTP Mix contains all four dNTPs each at a final concentration of 10 mM
Catalog No. 50-000-58944 Supplier Genscript Corporation Supplier No. C0158250
10 mM dNTP Mix is suitable for use in PCR sequencing nick translation fill-ins cDNA synthesis TdT tailing reactions and the dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in sterilized water The 10 mM dNTP Mix contains all four dNTPs each at a final concentration of 10 mM