Learn More
Genscript Corporation dNTP (25 mM each) ≥99% HPLC
Supplier:  Genscript Corporation C0158110
Encompass
25 mM dNTP Mix is suitable for use in PCR, sequencing, nick translation, fill-in, cDNA synthesis, TdT tailing reactions, and dilution of radiolabeled dNTPs. These nucleotides are supplied as convenient, ready-to-use solutions in sterilized water. The 25 mM dNTP Mix contains all four of the dNTPs, each at a final concentration of 25 mM.
Catalog No. 50-274-061