Learn More
Genscript Corporation dNTP 25 mM each 99 HPLC
Catalog No. 5000058349
Encompass
25 mM dNTP Mix is suitable for use in PCR sequencing nick translation fill-in cDNA synthesis TdT tailing reactions and dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in sterilized water The 25 mM dNTP Mix contains all four of the dNTPs each at a final concentration of 25 mM
Catalog No. 50-000-58349 Supplier Genscript Corporation Supplier No. C0158150
25 mM dNTP Mix is suitable for use in PCR sequencing nick translation fill-in cDNA synthesis TdT tailing reactions and dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in sterilized water The 25 mM dNTP Mix contains all four of the dNTPs each at a final concentration of 25 mM