Learn More
Meridian Life Science DNTP MASTERMIX 100MILLIMOLAR
SDP
Supplier:  Meridian Life Science BIO39028
Encompass
Ready to use molecular-grade dNTP mix containing the four dNTPs (dATP, dCTP, dGTP, dTTP). The mix is designed to save hands-on time for researchers and by reducing the amount of pipetting, makes contamination less likely, 100mM
Catalog No. NC9702419