Learn More
STEMCELL Technologies dNTP Mix (10 mM), Size: 1 mL
SDP
Catalog No. 502390358
Encompass
Simplify a variety of molecular biology processes by using dNTP Mix a solution that contains a mixture of four nucleotides (dATP dCTP dGTP dTTP). Each nucleotide is at a concentration of 10 mM in water at pH 7.5 and has greater than 99% purity. This dNTP Mix can be used in applications such as polymerase chain reaction (PCR) sequencing fill-in reactions and cDNA synthesis.
Catalog No. 50-239-0358 Supplier STEMCELL Technologies Supplier No. 76027
Simplify a variety of molecular biology processes by using dNTP Mix a solution that contains a mixture of four nucleotides (dATP dCTP dGTP dTTP). Each nucleotide is at a concentration of 10 mM in water at pH 7.5 and has greater than 99% purity. This dNTP Mix can be used in applications such as polymerase chain reaction (PCR) sequencing fill-in reactions and cDNA synthesis.