Learn More
Zymo Research Corporation dNTP Mix [10 mM] 100 ul
SDP
Catalog No. 501957579
Encompass
Highlights:- Ready to use dNTP Mix (dATP, dTTP, dGTP, dCTP) of ultrahigh purity; >99% trisphosphate by HPLC- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activities
Catalog No. 50-195-7579 Supplier Zymo Research Corporation Supplier No. D10001
Highlights:- Ready to use dNTP Mix (dATP, dTTP, dGTP, dCTP) of ultrahigh purity; >99% trisphosphate by HPLC- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activities