Learn More
Gold Biotechnology Inc dNTP Mix 10 x 1 mL
SDP
Catalog No. 5000562054
Encompass_Preferred
GoldBio s 2 -Deoxynucleoside 5 -triphosphate mix is an aqueous solution containing dATP dCTP dGTP and dTTP each at a final concentration of 10mM This mixture has been tested for performance in a wide size-range of PCR templates with Taq and Pfu DNA polymerases
Catalog No. 50-005-62054 Supplier Gold Biotechnology Inc Supplier No. D90010
GoldBio s 2 -Deoxynucleoside 5 -triphosphate mix is an aqueous solution containing dATP dCTP dGTP and dTTP each at a final concentration of 10mM This mixture has been tested for performance in a wide size-range of PCR templates with Taq and Pfu DNA polymerases