Learn More
Meridian Life Science DNTP MIX 1ML 10UM
SDP
Catalog No. NC9591361
Encompass
Ready to use molecular-grade dNTP mix containing the four dNTPs (dATP, dCTP, dGTP, dTTP). The mix is designed to save hands-on time for researchers and by reducing the amount of pipetting, makes contamination less likely, 100mM.
Catalog No. NC9591361 Supplier Meridian Life Science Supplier No. BIO39044
Ready to use molecular-grade dNTP mix containing the four dNTPs (dATP, dCTP, dGTP, dTTP). The mix is designed to save hands-on time for researchers and by reducing the amount of pipetting, makes contamination less likely, 100mM.