Learn More
Biotium dNTP Mix, 25mM each
Catalog No. 501964650
Encompass_Preferred
Highly purified mixes of dNTPs (>99% by HPLC) in 0.6 mM Tris-HCl (pH 7.5); each dNTP is at a concentration of 25 mM in the solution provided. The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.
Catalog No. 50-196-4650 Supplier Biotium Supplier No. 40053
Highly purified mixes of dNTPs (>99% by HPLC) in 0.6 mM Tris-HCl (pH 7.5); each dNTP is at a concentration of 25 mM in the solution provided. The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.