Learn More
As One International Inc dNTP mix, 25 mM of each dA, dC, dG and dT, 2 x 500 ul
Catalog No. 501656013
Encompass_Preferred
Mix of dATP, dCTP, dGTP, dTTP. Each nucleotide at a concentration of 25 mM.
Catalog No. 50-165-6013 Supplier As One International Inc Supplier No. AO500004
Mix of dATP, dCTP, dGTP, dTTP. Each nucleotide at a concentration of 25 mM.