Learn More
Apexbio Technology LLC 10 mM dNTP Mixture 10x1ml
SDP
Catalog No. 503107100
Encompass_Preferred
10 mM dNTP (2 -deoxynucleoside 5 -triphosphate) Mixture is an equimolar premixed aqueous solution containing four nucleotides (dATP dCTP dGTP and dTTP) each at a concentration of 10 mM The product is prepared as an aqueous solution titrated with NaOH to a neutral pH of 7 0
Catalog No. 50-310-7100 Supplier Apexbio Technology LLC Supplier No. K10415
10 mM dNTP (2 -deoxynucleoside 5 -triphosphate) Mixture is an equimolar premixed aqueous solution containing four nucleotides (dATP dCTP dGTP and dTTP) each at a concentration of 10 mM The product is prepared as an aqueous solution titrated with NaOH to a neutral pH of 7 0