Learn More
Takara Bio dNTP Mixture, 3.2 µmol Each/1.25 ml
SDP
Catalog No. 50443824
Encompass
This product is a mixture of dATP, dCTP, dGTP, and dTTP (each 2.5 mM). 3.2 µmol Each/1.25 ml
Catalog No. 50-443-824 Supplier Takara Bio Supplier No. 4030
This product is a mixture of dATP, dCTP, dGTP, and dTTP (each 2.5 mM). 3.2 µmol Each/1.25 ml