Learn More
Crescent Chemical Co Inc DNTP PCR MIX SOLUTION 10 MM
SDP
Catalog No. 50248419
Encompass_Preferred
dNTP PCR Mix, solution 10 mM molecular biology grade Ready to use dNTP mixture for the Tth DNA polymerase and Taq DNA polymerase.Mixture of dATP, dCTP, dGTP, dTTP, 10 mM of each, free of DNase, RNase, Phosphatase and Protease. Absolutely free of substances that may inhibit PCR
Catalog No. 50-248-419 Supplier Crescent Chemical Co Inc Supplier No. SE39712.02
dNTP PCR Mix, solution 10 mM molecular biology grade Ready to use dNTP mixture for the Tth DNA polymerase and Taq DNA polymerase.Mixture of dATP, dCTP, dGTP, dTTP, 10 mM of each, free of DNase, RNase, Phosphatase and Protease. Absolutely free of substances that may inhibit PCR