Learn More
Biotium dNTP Set, 100mM each
Catalog No. 501964649
Encompass_Preferred
Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mM. The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.
Catalog No. 50-196-4649 Supplier Biotium Supplier No. 40052
Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mM. The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.