Learn More
Thomas Scientific dNTP set dA, dG, dC, & dT, 100mM, 4x250æl/vial
SDP
Catalog No. 501459145
Encompass_Preferred
dNTP set dA, dG, dC, & dT 100mM 4x250ul/vial Storage Condition: -20ºC constant temperature freezer. Purity: Each dNTP is >99% chromatographically pure. Free of DNase, RNase, Protease, or Phosphatase activities. Application: For direct in-vitro synthesis of DNA or other related applications
Catalog No. 50-145-9145 Supplier Thomas Scientific Supplier No. C788T70
dNTP set dA, dG, dC, & dT 100mM 4x250ul/vial Storage Condition: -20ºC constant temperature freezer. Purity: Each dNTP is >99% chromatographically pure. Free of DNase, RNase, Protease, or Phosphatase activities. Application: For direct in-vitro synthesis of DNA or other related applications