Learn More
LAMDA BIOTECH INC dNTPs
Catalog No. NC3726051
Encompass_Preferred
A set of purified deoxynucleotide triphosphates (dATP, dCTP, dGTP, dTTP) for nucleic acid synthesis applications, tested for nuclease and phosphatase contamination.
Catalog No. NC3726051 Supplier LAMDA BIOTECH INC Supplier No. D107
A set of purified deoxynucleotide triphosphates (dATP, dCTP, dGTP, dTTP) for nucleic acid synthesis applications, tested for nuclease and phosphatase contamination.