Learn More
Qiagen Beverly Inc DNTP SOLUTION MIX
Catalog No. 50305950
Encompass
An Equimolar mixture of each dNTP (dATP, dCTP, dGTP, dTTP) at 25 mM and pH 7.5
Catalog No. 50-305-950 Supplier Qiagen Beverly Inc Supplier No. N2050L
An Equimolar mixture of each dNTP (dATP, dCTP, dGTP, dTTP) at 25 mM and pH 7.5