Learn More
Qiagen Beverly Inc DNTP SOLUTION SET
Catalog No. 50305951
Encompass
A set of four concentrated 100mM solutions of the four natural deoxynucleotides.
Catalog No. 50-305-951 Supplier Qiagen Beverly Inc Supplier No. N2010L
A set of four concentrated 100mM solutions of the four natural deoxynucleotides.