Learn More
LAMDA BIOTECH INC dNTPs
Supplier:  LAMDA BIOTECH INC D107L
Encompass_Preferred
Four tubes with 1000µl each of deoxyribonucleotide solution (dATP / dCTP / dGTP / dTTP) at 100mM in sterile, doubly distilled water. Final sodium concentration is 100mM. Dilute nucleotides in sterile, doubly distilled water.
Catalog No. 50-204-5773