Learn More
Crescent Chemical Co Inc DNTPS SET SOLUTION 100 MM
SDP
Catalog No. 50248415
Encompass_Preferred
dNTPs Set, solution 100 mM molecular biology grade Set of 100 mM solutions of each dATP, dCTP, dGTP, dTTP.Free of DNase, RNase, Phosphatase and Protease. Absolutely free of substances that may inhibit PCR, e.g. pyrophosphates etc.Ultrapure dNTPs qualified for use in standar
Catalog No. 50-248-415 Supplier Crescent Chemical Co Inc Supplier No. SE39705.02
dNTPs Set, solution 100 mM molecular biology grade Set of 100 mM solutions of each dATP, dCTP, dGTP, dTTP.Free of DNase, RNase, Phosphatase and Protease. Absolutely free of substances that may inhibit PCR, e.g. pyrophosphates etc.Ultrapure dNTPs qualified for use in standar