Learn More
Cayman Chemical Dantrolene-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2749234-17-9, 1 mg
SDP
Catalog No. 502991679
Encompass_Preferred
An internal standard for the quantification of dantrolene by GC- or LC-MS
Catalog No. 50-299-1679 Supplier Cayman Chemical Supplier No. 294511 MG
An internal standard for the quantification of dantrolene by GC- or LC-MS