Learn More
Aviva Systems Biology Corp DNTTIP1 Antibody - N-terminal region (ARP50740_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50740_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DNTTIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DNTTIP1. Alternate names: RP3-447F3.5, C20orf167, Tdif1, dJ447F3.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60423