Learn More
Medchemexpress LLC HY-114157 10mg Medchemexpress, DO-264 CAS:2301866-59-9 Purity:>98%
SDP
Catalog No. 501969293
Encompass_Preferred
Medchemexpress, HY-114157 10mg DO-264 CAS:2301866-59-9 DO-264 is a selective and in vivo-active inhibitor of Abhydrolase Domain Containing 12 (ABHD12), with an IC50 of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9293 Supplier Medchemexpress LLC Supplier No. HY11415710MG
Medchemexpress, HY-114157 10mg DO-264 CAS:2301866-59-9 DO-264 is a selective and in vivo-active inhibitor of Abhydrolase Domain Containing 12 (ABHD12), with an IC50 of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.