Learn More
MSC Dymon All-Purpose Cleaner, 18 oz Aerosol, Disinfectant, Odor-Free, 1 ea
Catalog No. 502866060
Encompass
This all-purpose cleaner is an 18 oz aerosol designed as a disinfectant with an odor-free scent, suitable for various hard and non-porous surfaces in institutional settings.
  • Aerosol form in a can
  • Container size: 18 oz
  • Odor-free scent
  • Disinfectant properties
  • EPA registered
  • Suitable for hospitals, hard and non-porous surfaces, and institutions
  • Effective on sinks, finished wood, glazed tile, glazed porcelain, and walls
  • Can be used on hampers, sink tops, chairs, fixtures, telephones, bathrooms, bathtubs, tables, cabinets, shower stalls, and bed springs
Catalog No. 50-286-6060 Supplier MSC Supplier No. 00322586
This all-purpose cleaner is an 18 oz aerosol designed as a disinfectant with an odor-free scent, suitable for various hard and non-porous surfaces in institutional settings.
  • Aerosol form in a can
  • Container size: 18 oz
  • Odor-free scent
  • Disinfectant properties
  • EPA registered
  • Suitable for hospitals, hard and non-porous surfaces, and institutions
  • Effective on sinks, finished wood, glazed tile, glazed porcelain, and walls
  • Can be used on hampers, sink tops, chairs, fixtures, telephones, bathrooms, bathtubs, tables, cabinets, shower stalls, and bed springs