Learn More
Broadley James Corporation Four Cartridge Kit for OxyProbe II
Catalog No. NC1537208
Encompass
This kit contains everything required to replace four membrane cartridges on any OxyProbe II sensor, packaged in one box for easy storage.
Catalog No. NC1537208 Supplier Broadley James Corporation Supplier No. MK2904
This kit contains everything required to replace four membrane cartridges on any OxyProbe II sensor, packaged in one box for easy storage.