Learn More
Bellco Glass, Inc. DO & pH Probe Holder Assembly For 18 Port
SDP
Catalog No. 501946417
Encompass_Preferred
pH/DO Probe Assy .or 18mm Port. To hold pH or DO 12mm Probes. Usage: 3,7,15L systems. MADE IN THE USA
Catalog No. 50-194-6417 Supplier Bellco Glass, Inc. Supplier No. 790900001
pH/DO Probe Assy .or 18mm Port. To hold pH or DO 12mm Probes. Usage: 3,7,15L systems. MADE IN THE USA