Learn More
Hach Company DO Probe Body Gasket
Supplier:  Hach Company 5797700
Encompass
Gasket-Dissolved Oxygen Probe BodyGasket used on Hach LDO Process Probe between probe body and sensor cap
Catalog No. 50-002-07208