Learn More
Bellco Glass, Inc. DO Probe PTFE,12X220mm Mettler
SDP
Catalog No. 501946318
Encompass_Preferred
Cathode platinumAnode silverPolarization 675mvOutput 40 to 70na in air at 25cTemperature 0 to 60c sterilization to 130cStability 2 or readout per week at constant pressure temp 30cLinearity better than 03 of readoutPressure 60psi maximumConnector 4 pin Bendix--Material welded parts 316L stainless steel
Catalog No. 50-194-6318 Supplier Bellco Glass, Inc. Supplier No. 780331545
Cathode platinumAnode silverPolarization 675mvOutput 40 to 70na in air at 25cTemperature 0 to 60c sterilization to 130cStability 2 or readout per week at constant pressure temp 30cLinearity better than 03 of readoutPressure 60psi maximumConnector 4 pin Bendix--Material welded parts 316L stainless steel