Learn More
Bellco Glass, Inc. DO Probe VP Adapter
SDP
Catalog No. 501946330
Encompass_Preferred
Straight adapter to connect a 4 pin style socket cable to a vp (variopin) probe connector.T82 style socket (pt02a-4p) for the cable
Catalog No. 50-194-6330 Supplier Bellco Glass, Inc. Supplier No. 780332000
Straight adapter to connect a 4 pin style socket cable to a vp (variopin) probe connector.T82 style socket (pt02a-4p) for the cable