Learn More
Bellco Glass, Inc. DO Probe W/VP,12x225mm With VP Connection - Ingold
SDP
Catalog No. 501946332
Encompass_Preferred
Measurement principle: amperometric / polarographicPressure resistance measurement: 0.2 – 6 bar absoluteMechanical pressure resistance: max. 12 bar (174.0 psi) absoluteMeasuring temperature range: 0 – 80°c (32 – 176°f)--Temperature range: -5 – 140°C (23 – 284°F)-- Temperature compensation: automatic with builtin RTD--Cable connection: vario pin (ip68)-- O-Ring material: Viton, silicone (FDA approved)-- Membrane material: Teflon / silicone / Teflon (reinforced with steel mesh)--Quick disconnect interior body: standard--Cathode: pt--Anode: ag--Guard ring: no--Detection limit: 6ppb--Accuracy: 1% or 4ppb--Response time @25°C (air to n2): 98% of--final value <90s-- Sensor signal in ambient air (25°C): 40 – 110na-- Residual signal in oxygen-free medium: <0.1% of the signal in ambient air.--Max. Flow error: less than or equal to: 5%-- Certification: EHEDG, 3a, 3.1b (en 10204.3.1b)
Catalog No. 50-194-6332 Supplier Bellco Glass, Inc. Supplier No. 780332225
Measurement principle: amperometric / polarographicPressure resistance measurement: 0.2 – 6 bar absoluteMechanical pressure resistance: max. 12 bar (174.0 psi) absoluteMeasuring temperature range: 0 – 80°c (32 – 176°f)--Temperature range: -5 – 140°C (23 – 284°F)-- Temperature compensation: automatic with builtin RTD--Cable connection: vario pin (ip68)-- O-Ring material: Viton, silicone (FDA approved)-- Membrane material: Teflon / silicone / Teflon (reinforced with steel mesh)--Quick disconnect interior body: standard--Cathode: pt--Anode: ag--Guard ring: no--Detection limit: 6ppb--Accuracy: 1% or 4ppb--Response time @25°C (air to n2): 98% of--final value <90s-- Sensor signal in ambient air (25°C): 40 – 110na-- Residual signal in oxygen-free medium: <0.1% of the signal in ambient air.--Max. Flow error: less than or equal to: 5%-- Certification: EHEDG, 3a, 3.1b (en 10204.3.1b)