Learn More
Chemglass Life Sciences DO Probe, w/VP, 12 x 225mm
SDP
Catalog No. 50143146
Encompass
DO Probe w/VP, 12 x 225mm. • Cathode: Platinum • Anode: Silver • Polarization: ~675mV • Output: 40 to 70nA in air at 25°C • Temperature: 0 to 60°C; Sterilization to 130°C • Stability: 2% or Readout per week at constant pressure/temp 30°C • Linearity: Better than ±0.3% of readout • Pressure: 60PSI maximum • Material: Wetted parts 316L SST • O-Ring: Viton
Catalog No. 50-143-146 Supplier Chemglass Life Sciences Supplier No. CLS1435125
DO Probe w/VP, 12 x 225mm. • Cathode: Platinum • Anode: Silver • Polarization: ~675mV • Output: 40 to 70nA in air at 25°C • Temperature: 0 to 60°C; Sterilization to 130°C • Stability: 2% or Readout per week at constant pressure/temp 30°C • Linearity: Better than ±0.3% of readout • Pressure: 60PSI maximum • Material: Wetted parts 316L SST • O-Ring: Viton