Learn More
METTLER TOLEDO DO Sensor InPro6860i/12/320/nA/HD
Catalog No. NC1807652 Shop All METTLER TOLEDO Products
Encompass
The InPro 6860i/12/120/mA/HD is an optical dissolved oxygen sensor with a mechanically modified hydrophilic surface. Combined with its tilted design, it prevents bubble accumulation, creating a highly stable measurement signal.
Catalog No. NC1807652 Supplier METTLER TOLEDO Supplier No. 30526901
The InPro 6860i/12/120/mA/HD is an optical dissolved oxygen sensor with a mechanically modified hydrophilic surface. Combined with its tilted design, it prevents bubble accumulation, creating a highly stable measurement signal.