Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DO2a-tert-butyl ester | 162148-48-3 | MFCD09263303 | >95.0% | 400.56 g/mol | C20H40N4O4 | 25 G
SDP

Catalog No. 5000282503
Encompass_Preferred

DO2A-tert-butyl ester is a bifunctional chelator used to conjugate peptides and radionuclides for the synthesis of imaging tracers and biochemical assay reagents. The compound has formula C20H40N4O4, molecular weight 400.56 g/mol, and CAS number 162148-48-3; storage recommendations include 4°C protected from light and, in solvent, -80°C (6 months) or -20°C (1 month).

  • Bifunctional chelator suitable for radionuclide conjugation.
  • Enables peptide coupling and radiolabeling workflows.
  • Applicable to development of radionuclide imaging tracers.
  • Chemical formula and molecular weight provided for formulation and calculation.
  • Storage and stability guidance available for solid and solution handling.
  • Purity varies by vendor; confirm batch certificate of analysis before use.

Catalog No. 50-002-82503 Supplier Medchemexpress LLC Supplier No. HYW28355625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DO2A-tert-butyl ester is a bifunctional chelator used to conjugate peptides and radionuclides for the synthesis of imaging tracers and biochemical assay reagents. The compound has formula C20H40N4O4, molecular weight 400.56 g/mol, and CAS number 162148-48-3; storage recommendations include 4°C protected from light and, in solvent, -80°C (6 months) or -20°C (1 month).

  • Bifunctional chelator suitable for radionuclide conjugation.
  • Enables peptide coupling and radiolabeling workflows.
  • Applicable to development of radionuclide imaging tracers.
  • Chemical formula and molecular weight provided for formulation and calculation.
  • Storage and stability guidance available for solid and solution handling.
  • Purity varies by vendor; confirm batch certificate of analysis before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.