Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DO2A-tert-butyl ester | 162148-48-3 | MFCD09263303 | 98.0% | 400.56 | C20H40N4O4 | 10 G
SDP

Catalog No. 5000281317
Encompass_Preferred

DO2A-tert-butyl ester is a bifunctional chelator intended for conjugation of peptides and radionuclides in radiolabeling and bioconjugation workflows. It contains diacetate chelating groups that enable stable metal coordination and is supplied with solvent preparation and storage guidance for research applications.

  • Bifunctional chelator for peptide and radionuclide coupling.
  • Enables stable coordination of metal ions during radiolabeling.
  • Solvent preparation volumes provided for common sample sizes.
  • Storage recommendations included for short- and long-term handling.
  • Suitable for research use in bioconjugation workflows.

Catalog No. 50-002-81317 Supplier Medchemexpress LLC Supplier No. HYW28355610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DO2A-tert-butyl ester is a bifunctional chelator intended for conjugation of peptides and radionuclides in radiolabeling and bioconjugation workflows. It contains diacetate chelating groups that enable stable metal coordination and is supplied with solvent preparation and storage guidance for research applications.

  • Bifunctional chelator for peptide and radionuclide coupling.
  • Enables stable coordination of metal ions during radiolabeling.
  • Solvent preparation volumes provided for common sample sizes.
  • Storage recommendations included for short- and long-term handling.
  • Suitable for research use in bioconjugation workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.