Learn More
Medchemexpress LLC HY-117771 10mg Medchemexpress, DO34 CAS:1848233-58-8 Purity:>98%
SDP
Catalog No. 501969490
Encompass_Preferred
Medchemexpress, HY-117771 10mg DO34 CAS:1848233-58-8 DO34 is a highly potent, selective and centrally active diacylglycerol lipase (DAGL) inhibitor, with an IC50 of 6 nM for DAGLα conversion of SAG to 2-AG, and an IC50 for DAGLβ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9490 Supplier Medchemexpress LLC Supplier No. HY11777110MG
Medchemexpress, HY-117771 10mg DO34 CAS:1848233-58-8 DO34 is a highly potent, selective and centrally active diacylglycerol lipase (DAGL) inhibitor, with an IC50 of 6 nM for DAGLα conversion of SAG to 2-AG, and an IC50 for DAGLβ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.