Learn More
Medchemexpress LLC DO3A tert-Butyl ester | 122555-91-3 | 98.0% | 5 G
SDP
Catalog No. 5000340890
Encompass_Preferred
DO3A tert-Butyl ester is a cyclic tosamide benzyl derivative intended for research use only. It exhibits cytotoxicity in various cancer cells.
  • Intended for research use only
  • Exhibits cytotoxicity in cancer cells A375, A2780, HT29, MCF-7, FaDu, and NIH 3T3
  • IC50 values ranging from 8.7 to 17.3 μM
Catalog No. 50-003-40890 Supplier Medchemexpress LLC Supplier No. HY126925G
DO3A tert-Butyl ester is a cyclic tosamide benzyl derivative intended for research use only. It exhibits cytotoxicity in various cancer cells.
  • Intended for research use only
  • Exhibits cytotoxicity in cancer cells A375, A2780, HT29, MCF-7, FaDu, and NIH 3T3
  • IC50 values ranging from 8.7 to 17.3 μM