Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories DOA Positive Control

Catalog No. 23111316
Change view
Click to view available options
Quantity:
1 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-316 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-316 Supplier Germaine™ Laboratories Supplier No. 40002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories AimScreen™ DOA Controls monitor and validate performance of DOA detection methods at levels established by SAMHSA, CAP/AACC and many state programs

  • Compatible with all quantitative and qualitative drug detection procedures which are sufficiently sensitive to detect the control constituents
  • Positive control urines have been spiked with authentic reference drug standards and/or appropriate metabolites
  • Should be treated as any unknown specimen while following the specific protocol of the assay being used
  • Controls prepared from human based urine available as a negative
  • Cutoff is +3x to monitor the performance of drugs of abuse test devices
  • Each bottle contains stabilized human based urine
  • Human urine based matrix

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes AMP, BAR, Buprenorphine, BZO, COC, MDMA, mAMP, MOP, MTD, OPI, Oxycodone, PCP, PPx, TCA, THC
For Use With (Equipment) All quantitative and qualitative drug detection procedures which are sufficiently sensitive to detect the control constituents.
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.