Learn More
Medchemexpress LLC HY-15746 100mg Medchemexpress, Dobutamine (hydrochloride) CAS:49745-95-1 Purity:>98%
SDP
Catalog No. 502081788
Encompass_Preferred
Medchemexpress, HY-15746 100mg Dobutamine (hydrochloride) CAS:49745-95-1 Dobutamine hydrochloride is a synthetic catecholamine that acts on α1-AR, β1-AR, β2-AR (α-1, β-1 andβ-2 adrenoceptors). Dobutamine hydrochloride is a selective β1-AR agonist, relatively weak activity at α1-AR and β2-AR. Dobutamine hydrochloride can increase cardiac output and correct hypoperfusion[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1788 Supplier Medchemexpress LLC Supplier No. HY15746100MG
Medchemexpress, HY-15746 100mg Dobutamine (hydrochloride) CAS:49745-95-1 Dobutamine hydrochloride is a synthetic catecholamine that acts on α1-AR, β1-AR, β2-AR (α-1, β-1 andβ-2 adrenoceptors). Dobutamine hydrochloride is a selective β1-AR agonist, relatively weak activity at α1-AR and β2-AR. Dobutamine hydrochloride can increase cardiac output and correct hypoperfusion[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.