Learn More
Cayman Chemical DobutamInhydrochlorIde 10mg
SDP
Catalog No. 502972066
Encompass_Preferred
A β-adrenergic receptor agonist with potent positive inotropic effects in vivo; strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions; strongly increases cardiac contractility
Catalog No. 50-297-2066 Supplier Cayman Chemical Supplier No. 1558210 MG
A β-adrenergic receptor agonist with potent positive inotropic effects in vivo; strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions; strongly increases cardiac contractility