Learn More
Cayman Chemical Dobutamine (hydrochloride), Purity: ≥95%, CAS: 49745-95-1, 50 mg
SDP
Catalog No. 502968906
Encompass_Preferred
A β-adrenergic receptor agonist with potent positive inotropic effects in vivo; strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions; strongly increases cardiac contractility
Catalog No. 50-296-8906 Supplier Cayman Chemical Supplier No. 1558250 MG
A β-adrenergic receptor agonist with potent positive inotropic effects in vivo; strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions; strongly increases cardiac contractility