Learn More
MSC Value Collection Packing Slip Envelope, 10 x 6.5 in, 1000/Pk
Catalog No. 501920787
Encompass
Pressure sensitive packing list envelopes secure and protect documents attached to the outside of shipments. They are constructed from water-resistant heavy 2 mil poly and feature a hot melt adhesive backing for strong adhesion.
  • Excellent protection and security: keeps paperwork protected and secure in transit.
  • Weather resistant: durable polyethylene protects important documents from dirt, moisture, and the elements.
  • Versatile and easy to use: can be applied to boxes, envelopes, mailing bags, tubes, and more with permanent adhesive.
Catalog No. 50-192-0787 Supplier MSC Supplier No. 39520614
Pressure sensitive packing list envelopes secure and protect documents attached to the outside of shipments. They are constructed from water-resistant heavy 2 mil poly and feature a hot melt adhesive backing for strong adhesion.
  • Excellent protection and security: keeps paperwork protected and secure in transit.
  • Weather resistant: durable polyethylene protects important documents from dirt, moisture, and the elements.
  • Versatile and easy to use: can be applied to boxes, envelopes, mailing bags, tubes, and more with permanent adhesive.