Learn More
Medchemexpress LLC HY-12886 5mg Medchemexpress, Docebenone CAS:80809-81-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128865MG
Encompass_Preferred
Medchemexpress, HY-12886 5mg Docebenone CAS:80809-81-0 Docebenone (AA 861) is a potent, selective and orally active 5-LO (5-lipoxygenase) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0138