Learn More
Apexbio Technology LLC Docetaxel 114977-28-5 100mg
SDP
Catalog No. 5000062458
Encompass_Preferred
Docetaxel (CAS 114977-28-5) is a semisynthetic taxane derivative originally isolated from the European yew (Taxus baccata) As an antineoplastic agent docetaxel disrupts microtubule dynamics by stabilizing tubulin polymerization resulting in cell cycle arrest at mitosis and subsequent apoptosis In both in vitro and in vivo studies docetaxel demonstrates pronounced cytotoxic activity against diverse tumor types such as breast lung ovarian head and neck and gastric cancers Research particularly highlights its enhanced potency against ovarian cancer cell lines compared to paclitaxel cisplatin and etoposide indicating substantial research value in oncology programs
Catalog No. 50-000-62458 Supplier Apexbio Technology LLC Supplier No. A4394100
Docetaxel (CAS 114977-28-5) is a semisynthetic taxane derivative originally isolated from the European yew (Taxus baccata) As an antineoplastic agent docetaxel disrupts microtubule dynamics by stabilizing tubulin polymerization resulting in cell cycle arrest at mitosis and subsequent apoptosis In both in vitro and in vivo studies docetaxel demonstrates pronounced cytotoxic activity against diverse tumor types such as breast lung ovarian head and neck and gastric cancers Research particularly highlights its enhanced potency against ovarian cancer cell lines compared to paclitaxel cisplatin and etoposide indicating substantial research value in oncology programs