Learn More
Medchemexpress LLC Docetaxel | 114977-28-5 | 99.9% | C43H53NO14 | 1 ML
SDP
Catalog No. 5000432508
Encompass_Preferred
Docetaxel is a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, leading to cell cycle arrest at G2/M and subsequent apoptosis. This compound demonstrates anti-cancer activity.
  • Microtubule depolymerization inhibitor
  • Attenuates effects of bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Leads to cell apoptosis
  • Demonstrates anti-cancer activity
Catalog No. 50-004-32508 Supplier Medchemexpress LLC Supplier No. HYB001110MM/1ML
Docetaxel is a microtubule depolymerization inhibitor with an IC50 of 0.2 μM. It attenuates the effects of bcl-2 and bcl-xL gene expression, leading to cell cycle arrest at G2/M and subsequent apoptosis. This compound demonstrates anti-cancer activity.
  • Microtubule depolymerization inhibitor
  • Attenuates effects of bcl-2 and bcl-xL gene expression
  • Arrests cell cycle at G2/M
  • Leads to cell apoptosis
  • Demonstrates anti-cancer activity