Learn More
Apexbio Technology LLC Docetaxel 114977-28-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061852
Encompass_Preferred
Docetaxel (CAS 114977-28-5) is a semisynthetic taxane derivative originally isolated from the European yew (Taxus baccata) As an antineoplastic agent docetaxel disrupts microtubule dynamics by stabilizing tubulin polymerization resulting in cell cycle arrest at mitosis and subsequent apoptosis In both in vitro and in vivo studies docetaxel demonstrates pronounced cytotoxic activity against diverse tumor types such as breast lung ovarian head and neck and gastric cancers Research particularly highlights its enhanced potency against ovarian cancer cell lines compared to paclitaxel cisplatin and etoposide indicating substantial research value in oncology programs
Catalog No. 50-000-61852 Supplier Apexbio Technology LLC Supplier No. A43945.1
Docetaxel (CAS 114977-28-5) is a semisynthetic taxane derivative originally isolated from the European yew (Taxus baccata) As an antineoplastic agent docetaxel disrupts microtubule dynamics by stabilizing tubulin polymerization resulting in cell cycle arrest at mitosis and subsequent apoptosis In both in vitro and in vivo studies docetaxel demonstrates pronounced cytotoxic activity against diverse tumor types such as breast lung ovarian head and neck and gastric cancers Research particularly highlights its enhanced potency against ovarian cancer cell lines compared to paclitaxel cisplatin and etoposide indicating substantial research value in oncology programs