Learn More
AdipoGen Docetaxel
SDP
Catalog No. 502561098
Encompass_Preferred
Chemical. CAS 114977-28-5. Formula C43H53NO14. MW 807.88. Synthetic. Anticancer agent. Semisynthetic analog of taxol paclitaxel. Inhibits microtubule disassembly and inhibits cell replication. Binds to and stabilizes the beta−tubulin subunit of microtubules, preventing depolymerization of the mitotic spindle thus leading to cell cycle arrest at G2/M and apoptosis. This cytotoxic activity has been widely exploited in suppressing oncogenic proliferation. Displays potent and broad antineoplastic properties, used mainly for the treatment of breast, ovarian, and non-small cell lung cancer. Also inhibits pro-angiogenic factors such as vascular endothelial growth factor VEGF and displays immunomodulatory and pro-inflammatory properties by inducing various mediators of the inflammatory response. Also studied for use as a radiation-sensitizing agent.
Catalog No. 50-256-1098 Supplier AdipoGen Supplier No. CDXD0371M025
Chemical. CAS 114977-28-5. Formula C43H53NO14. MW 807.88. Synthetic. Anticancer agent. Semisynthetic analog of taxol paclitaxel. Inhibits microtubule disassembly and inhibits cell replication. Binds to and stabilizes the beta−tubulin subunit of microtubules, preventing depolymerization of the mitotic spindle thus leading to cell cycle arrest at G2/M and apoptosis. This cytotoxic activity has been widely exploited in suppressing oncogenic proliferation. Displays potent and broad antineoplastic properties, used mainly for the treatment of breast, ovarian, and non-small cell lung cancer. Also inhibits pro-angiogenic factors such as vascular endothelial growth factor VEGF and displays immunomodulatory and pro-inflammatory properties by inducing various mediators of the inflammatory response. Also studied for use as a radiation-sensitizing agent.