Learn More
Medchemexpress LLC HY-B0011 200mg , Docetaxel CAS:114977-28-5 Purity:98%
SDP
Catalog No. 501492528
Encompass_Preferred
Medchemexpress, HY-B0011 200mg Docetaxel CAS:114977-28-5 Formula:C43H53NO14 Microtubule Purity:98% Docetaxel is a semi-synthetic taxane analogue, acts as a microtubule stabilizer. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2528 Supplier Medchemexpress LLC Supplier No. HYB0011200MG
Medchemexpress, HY-B0011 200mg Docetaxel CAS:114977-28-5 Formula:C43H53NO14 Microtubule Purity:98% Docetaxel is a semi-synthetic taxane analogue, acts as a microtubule stabilizer. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.