Learn More
TARGETMOL CHEMICALS INC Docetaxel 500MG
SDP
Catalog No. 502880614
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Docetaxel (RP-56976), a semi-synthetic analog of paclitaxel, is a microtubule depolymerization inhibitor (IC50=0.2 uM) that attenuates the effects of bcl-2 and bcl-xL gene expression and exhibits apoptosis-inducing, anti-tumor activity. Purity 99.1%
Catalog No. 50-288-0614 Supplier TARGETMOL CHEMICALS INC Supplier No. T1034500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Docetaxel (RP-56976), a semi-synthetic analog of paclitaxel, is a microtubule depolymerization inhibitor (IC50=0.2 uM) that attenuates the effects of bcl-2 and bcl-xL gene expression and exhibits apoptosis-inducing, anti-tumor activity. Purity 99.1%