Learn More
Selleck Chemical LLC Docetaxel S1148-250mg
SDP
Catalog No. 503108362
Encompass_Preferred
Docetaxel an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules
Catalog No. 50-310-8362 Supplier Selleck Chemical LLC Supplier No. S1148250MG
Docetaxel an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules