Learn More
Selleck Chemical LLC Docetaxel S1148-1g
SDP
Catalog No. 503111076
Encompass_Preferred
Docetaxel an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules
Catalog No. 50-311-1076 Supplier Selleck Chemical LLC Supplier No. S11481G
Docetaxel an analog of paclitaxel is an inhibitor of depolymerisation of microtubules by binding to stabilized microtubules